ENA100801908 Enamine

5-nitro-1,2,3,4-tetrahydropyridazine-3,4-dione

  • MDL number MFCD23146232
  • Cas Number : 2854-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25564
Availability details for
Quantity
  • Availability
  • Add to Cart