ENA100801910 Enamine

6-(2,3-dihydro-1,4-benzodioxin-6-yl)-1-isopropyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08691199
  • Cas Number : 924477-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26083

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26083
Availability details for
Quantity
  • Availability
  • Add to Cart