ENA100801911 Enamine

2-[(2-tert-butylphenoxy)methyl]oxirane

  • MDL number MFCD08444094
  • Cas Number : 40786-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...211
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22518
Availability details for
Quantity
  • Availability
  • Add to Cart