ENA100801926 Enamine

1-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00172902
  • Cas Number : 7465-66-9
  • Molecular Weight : 151.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26104
Availability details for
Quantity
  • Availability
  • Add to Cart