ENA100801928 Enamine

N-(6-amino-1-butyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)-N-benzyl-2-chloroacetamide

  • MDL number MFCD08444101
  • Cas Number : 923762-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22591
Availability details for
Quantity
  • Availability
  • Add to Cart