ENA100801929 Enamine

2-(chloromethyl)-5-ethyl-1,3,4-oxadiazole

  • MDL number MFCD08059898
  • Cas Number : 3914-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25580
Availability details for
Quantity
  • Availability
  • Add to Cart