ENA100801932 Enamine

2-chloro-1-[4-(3,4-dimethoxybenzenesulfonyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD05862826
  • Cas Number : 925407-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22631

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22631
Availability details for
Quantity
  • Availability
  • Add to Cart