ENA100801933 Enamine

4-chloro-3-nitro-N-(propan-2-yl)benzamide

  • MDL number MFCD01182366
  • Cas Number : 39887-54-2
  • Molecular Weight : 242.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25583
Availability details for
Quantity
  • Availability
  • Add to Cart