ENA100801936 Enamine

2-[(benzylamino)methyl]-3,4-dihydroquinazolin-4-one

  • MDL number MFCD07759152
  • Cas Number : 143993-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25586

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25586
Availability details for
Quantity
  • Availability
  • Add to Cart