ENA100801943 Enamine

(1-phenyl-1H-pyrazol-4-yl)methylamine

  • MDL number MFCD02656664
  • Cas Number : 400877-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26123

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-26123
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart