ENA102117485 Enamine

2-chloro-N-{2-[4-(difluoromethoxy)phenyl]ethyl}acetamide

  • MDL number MFCD08729195
  • Cas Number : 929973-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23260
Availability details for
Quantity
  • Availability
  • Add to Cart