ENA102117486 Enamine

2-chloro-N-[2-(4-ethoxyphenoxy)ethyl]acetamide

  • MDL number MFCD08729196
  • Cas Number : 929973-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23288
Availability details for
Quantity
  • Availability
  • Add to Cart