ENA102117487 Enamine

2-chloro-N-(4-ethyl-4-methyl-2,5-dioxoimidazolidin-1-yl)acetamide

  • MDL number MFCD08729197
  • Cas Number : 957011-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23298
Availability details for
Quantity
  • Availability
  • Add to Cart