ENA102117502 Enamine

4-[(3,5-dimethylpiperidin-1-yl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD08729213
  • Cas Number : 929974-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26575
Availability details for
Quantity
  • Availability
  • Add to Cart