ENA102117503 Enamine

4-[(2,6-dimethylmorpholin-4-yl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD08729214
  • Cas Number : 929974-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26576

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26576
Availability details for
Quantity
  • Availability
  • Add to Cart