ENA102117506 Enamine

7-ethyl-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole

  • MDL number MFCD08729217
  • Cas Number : 460354-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26583
Availability details for
Quantity
  • Availability
  • Add to Cart