ENA102117509 Enamine

1-[(2-chlorophenyl)methyl]-6-phenyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08729220
  • Cas Number : 929974-35-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26591
Availability details for
Quantity
  • Availability
  • Add to Cart