ENA102117510 Enamine

1-(2-furylmethyl)-6-phenyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08729222
  • Cas Number : 929974-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 242...244
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26594

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26594
Availability details for
Quantity
  • Availability
  • Add to Cart