ENA102117511 Enamine

6-(pyridin-3-yl)-1-[(pyridin-3-yl)methyl]-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08729223
  • Cas Number : 929974-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-26595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26595
Availability details for
Quantity
  • Availability
  • Add to Cart