ENA102117524 Enamine

3-({[(4-methoxyphenyl)amino]carbonyl}amino)propanoic acid

  • MDL number MFCD08729235
  • Cas Number : 929974-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26642

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26642
Availability details for
Quantity
  • Availability
  • Add to Cart