ENA102117525 Enamine

3-bromo-1-chloro-4-methoxy-2,5-dinitrobenzene

  • MDL number MFCD08729236
  • Cas Number : 929974-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26645
Availability details for
Quantity
  • Availability
  • Add to Cart