ENA102117526 Enamine

2,4-ditrifluoromethanesulfonylphenol

  • MDL number MFCD08729237
  • Cas Number : 15183-81-0
  • Molecular Weight : 358.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26648
Availability details for
Quantity
  • Availability
  • Add to Cart