ENA102117530 Enamine

4-[(diethylamino)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD08729241
  • Cas Number : 859632-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26655
Availability details for
Quantity
  • Availability
  • Add to Cart