ENA102117546 Enamine

1-{[(benzylamino)carbonyl]amino}cyclopentanecarboxylic acid

  • MDL number MFCD08729262
  • Cas Number : 929974-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26718

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26718
Availability details for
Quantity
  • Availability
  • Add to Cart