ENA102117550 Enamine

1-oxaspiro[2.6]nonane

  • MDL number MFCD08729226
  • Cas Number : 185-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26610

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26610
Availability details for
Quantity
  • Availability
  • Add to Cart