ENA102117555 Enamine

5-imino-4H,5H-[1,3]thiazolo[4,5-b]pyridin-2-amine

  • MDL number MFCD23145002
  • Cas Number : 13575-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193-194
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-16744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16744
Availability details for
Quantity
  • Availability
  • Add to Cart