ENA102117565 Enamine

1-(2-chloroacetyl)-N,N-dimethyl-1,2,3,4-tetrahydroquinoline-6-sulfonamide

  • MDL number MFCD08729201
  • Cas Number : 923794-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23815
Availability details for
Quantity
  • Availability
  • Add to Cart