ENA102117566 Enamine

4-{[(2-methoxyethyl)amino]sulfonyl}benzoic acid

  • MDL number MFCD05804385
  • Cas Number : 716358-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 254...256
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18357

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18357
Availability details for
Quantity
  • Availability
  • Add to Cart