ENA102117570 Enamine

2-bromo-4-chloro-6-nitrophenol

  • MDL number MFCD00024243
  • Cas Number : 15969-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126...128
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26580

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26580
Availability details for
Quantity
  • Availability
  • Add to Cart