ENA102117596 Enamine

3-acetamido-3-(thiophen-2-yl)propanoic acid

  • MDL number MFCD01084326
  • Cas Number : 382609-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26319
Availability details for
Quantity
  • Availability
  • Add to Cart