ENA102117597 Enamine

2-chloro-5-phenyl-1,3,4-thiadiazole

  • MDL number MFCD01109216
  • Cas Number : 13373-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26679
Availability details for
Quantity
  • Availability
  • Add to Cart