ENA102117601 Enamine

N-(3-cyanophenyl)-N'-phenylurea

  • MDL number MFCD02026281
  • Cas Number : 87597-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26685

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26685
Availability details for
Quantity
  • Availability
  • Add to Cart