ENA102117602 Enamine

4,5-dimethyl-1-(tetrahydrofuran-2-ylmethyl)-1H-imidazole-2-thiol

  • MDL number MFCD06386702
  • Cas Number : 851814-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18581

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18581
Availability details for
Quantity
  • Availability
  • Add to Cart