ENA102117610 Enamine

2-[5-(chloromethyl)-1,2,4-oxadiazol-3-yl]pyridine

  • MDL number MFCD08729256
  • Cas Number : 90002-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102...104
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26702

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26702
Availability details for
Quantity
  • Availability
  • Add to Cart