ENA102117611 Enamine

N-(4-cyanophenyl)-2-furamide

  • MDL number MFCD01815679
  • Cas Number : 332065-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26704

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26704
Availability details for
Quantity
  • Availability
  • Add to Cart