ENA102117617 Enamine

(3-methyl-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)acetic acid

  • MDL number MFCD00850016
  • Cas Number : 1848-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...204
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26603

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26603
Availability details for
Quantity
  • Availability
  • Add to Cart