ENA102117622 Enamine

6-bromo-2H-chromene-3-carboxylic acid

  • MDL number MFCD04035698
  • Cas Number : 380607-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243...245
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26717

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26717
Availability details for
Quantity
  • Availability
  • Add to Cart