ENA111667454 Enamine

2-chloro-N-{4-chloro-3-[(dimethylamino)sulfonyl]phenyl}propanamide

  • MDL number MFCD09040362
  • Cas Number : 929973-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23332

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23332
Availability details for
Quantity
  • Availability
  • Add to Cart