ENA111667463 Enamine

ethyl 2-(cyanomethyl)-4-methylquinoline-3-carboxylate

  • MDL number MFCD09040387
  • Cas Number : 929974-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26701
Availability details for
Quantity
  • Availability
  • Add to Cart