ENA111667472 Enamine

4-(N-methyl4-methoxybenzenesulfonamido)butanoic acid

  • MDL number MFCD09040400
  • Cas Number : 929975-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26749

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26749
Availability details for
Quantity
  • Availability
  • Add to Cart