ENA111667476 Enamine

4,4,4-trifluoro-3-hydroxy-3-(5-methyl-2-furyl)butanenitrile

  • MDL number MFCD09040404
  • Cas Number : 929975-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26772

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26772
Availability details for
Quantity
  • Availability
  • Add to Cart