ENA111667516 Enamine

1-isopropyl-4-methyl-1,7-dihydro-6H-pyrazolo[3,4-b]pyridin-6-one

  • MDL number MFCD09040398
  • Cas Number : 1934472-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26743

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26743
Availability details for
Quantity
  • Availability
  • Add to Cart