ENA111667519 Enamine

N-(3-aminophenyl)-N'-phenylurea

  • MDL number MFCD08532932
  • Cas Number : 66695-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26799

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26799
Availability details for
Quantity
  • Availability
  • Add to Cart