ENA111667520 Enamine

N-(4-aminophenyl)-2-morpholin-4-ylacetamide

  • MDL number MFCD03274587
  • Cas Number : 105076-76-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26744

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26744
Availability details for
Quantity
  • Availability
  • Add to Cart