ENA111667530 Enamine

ethyl 5-amino-1-(2-naphthyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD01918174
  • Cas Number : 1955498-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26813

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26813
Availability details for
Quantity
  • Availability
  • Add to Cart