ENA111667545 Enamine

2-mercapto-3-(3-methoxypropyl)pyrido[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD09040393
  • Cas Number : 688793-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26736

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26736
Availability details for
Quantity
  • Availability
  • Add to Cart