ENA112933047 Enamine

1-(1,3-benzothiazol-2-yl)-2,2,2-trifluoroethan-1-one

  • MDL number MFCD09040352
  • Cas Number : 929973-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23116
Availability details for
Quantity
  • Availability
  • Add to Cart