ENA112933053 Enamine

1-(2-chloroacetyl)-2-methyl-2,3-dihydro-1H-indole-5-sulfonamide

  • MDL number MFCD09040375
  • Cas Number : 929973-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-23819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23819
Availability details for
Quantity
  • Availability
  • Add to Cart