ENA112933054 Enamine

ethyl (2E)-3-{4-[(chloroacetyl)amino]phenyl}acrylate

  • MDL number MFCD09040377
  • Cas Number : 92026-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24610

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24610
Availability details for
Quantity
  • Availability
  • Add to Cart