ENA112933059 Enamine

8-nitro-3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonyl chloride

  • MDL number MFCD09040426
  • Cas Number : 929975-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26840
Availability details for
Quantity
  • Availability
  • Add to Cart